Learn More
Biolegend APC anti-mouse CD4
Supplier:  Biolegend 116013
Encompass_Preferred
APC anti-mouse CD4 [RM4-4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0226