Learn More
Biolegend APC anti-mouse CD41
Supplier:  Biolegend 133914
Encompass_Preferred
APC anti-mouse CD41 [MWReg30]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-712