Learn More
Biolegend APC anti-mouse/human CD45R/B220
Supplier:  Biolegend 103212
Encompass_Preferred
APC anti-mouse/human CD45R/B220 [RA3-6B2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cat (Feline); Apps: FC; Size: 100 ug
Catalog No. 50-162-804