Learn More
Biolegend APC anti-mouse CD62L
Supplier:  Biolegend 104411
Encompass_Preferred
APC anti-mouse CD62L [MEL-14]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-918