Learn More
Biolegend APC anti-mouse CD63
Supplier:  Biolegend 143905
Encompass_Preferred
APC anti-mouse CD63 [NVG-2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC; Size: 25 ug
Catalog No. 50-207-1477