Learn More
Biolegend APC anti-mouse CD66a (CEACAM1a)
Supplier:  Biolegend 134509
Encompass_Preferred
APC anti-mouse CD66a (CEACAM1a) [MAb-CC1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-1030