Learn More
Biolegend APC anti-mouse CD86
Supplier:  Biolegend 105114
Encompass_Preferred
APC anti-mouse CD86 [PO3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-025