Learn More
Biolegend APC anti-mouse CD90.2 (Thy1.2)
Supplier:  Biolegend 140311
Encompass_Preferred
APC anti-mouse CD90.2 (Thy1.2) [53-2.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-165-129