Learn More
Biolegend APC anti-mouse CD93 (AA4.1, early B lineage)
Supplier:  Biolegend 136510
Encompass_Preferred
APC anti-mouse CD93 (AA4.1, early B lineage) [AA4.1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-924