Learn More
Biolegend APC anti-mouse DC Marker (33D1)
Supplier:  Biolegend 124913
Encompass_Preferred
APC anti-mouse DC Marker (33D1) [33D1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-223