Learn More
Biolegend APC anti-mouse DLL1
Supplier:  Biolegend 128314
Encompass_Preferred
APC anti-mouse DLL1 [HMD1-3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-469