Learn More
Biolegend APC anti-mouse F4/80
Supplier:  Biolegend 123115
Encompass_Preferred
APC anti-mouse F4/80 [BM8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC9844144