Learn More
Biolegend APC anti-mouse H-2Kd
Supplier:  Biolegend 116619
Encompass_Preferred
APC anti-mouse H-2Kd [SF1-1.1]; Isotype: Mouse (SJL) IgG2a, κ; Reactivity: Mouse d haplotype, Weakly cross-reacts with H-2k haplotype; Apps: FC; Size: 25 ug
Catalog No. 50-163-692