Learn More
Biolegend APC anti-mouse Ig light chain λ
Supplier:  Biolegend 407306
Encompass_Preferred
APC anti-mouse Ig light chain λ [RML-42]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 ug
Catalog No. 50-169-198