Learn More
Biolegend APC anti-mouse IgD
Supplier:  Biolegend 405714
Encompass_Preferred
APC anti-mouse IgD [11-26c.2a]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-169-128