Learn More
Biolegend APC anti-mouse IL-10
Supplier:  Biolegend 505009
Encompass_Preferred
APC anti-mouse IL-10 [JES5-16E3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 ug
Catalog No. 50-169-877