Learn More
Biolegend APC anti-mouse IL-21R
Supplier:  Biolegend 131909
Encompass_Preferred
APC anti-mouse IL-21R [4A9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-639