Learn More
Biolegend APC anti-mouse IL-27 p28
Supplier:  Biolegend 516906
Encompass_Preferred
APC anti-mouse IL-27 p28 [MM27-7B1]; Isotype: Mouse IgG2a; Reactivity: Mouse, Human; Apps: ICFC; Size: 100 ug
Catalog No. 50-170-211