Learn More
Biolegend APC anti-mouse LPAM-1 (Integrin α4β7)
Supplier:  Biolegend 120608
Encompass_Preferred
APC anti-mouse LPAM-1 (Integrin α4β7) [DATK32]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-869