Learn More
Biolegend APC anti-mouse Ly-6A/E (Sca-1)
Supplier:  Biolegend 108112
Encompass_Preferred
APC anti-mouse Ly-6A/E (Sca-1) [D7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-217