Learn More
Biolegend APC anti-mouse Ly-6G
Supplier:  Biolegend 127613
Encompass_Preferred
APC anti-mouse Ly-6G [1A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-394