Learn More
Biolegend APC anti-mouse Notch 2
Supplier:  Biolegend 130713
Encompass_Preferred
APC anti-mouse Notch 2 [HMN2-35]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-591