Learn More
Biolegend APC anti-mouse TCR γ/δ
Supplier:  Biolegend 118116
Encompass_Preferred
APC anti-mouse TCR γ/δ [GL3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC9083256