Learn More
Biolegend APC anti-mouse TCR Vγ2
Supplier:  Biolegend 137707
Encompass_Preferred
APC anti-mouse TCR Vγ2 [UC3-10A6]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-992