Learn More
Biolegend APC anti-mouse TIGIT (Vstm3)
Supplier:  Biolegend 142105
Encompass_Preferred
APC anti-mouse TIGIT (Vstm3) [1G9]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-165-246