Learn More
Biolegend APC anti-mouse CD366 (Tim-3)
Supplier:  Biolegend 119705
Encompass_Preferred
APC anti-mouse CD366 (Tim-3) [RMT3-23]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-604-940