Learn More
Biolegend APC anti-mouse TCR Vα2
Supplier:  Biolegend 127809
Encompass_Preferred
APC anti-mouse TCR Vα2 [B20.1]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-413