Learn More
Biolegend APC anti-rat CD71
Supplier:  Biolegend 204411
Encompass_Preferred
APC anti-rat CD71 [OX-26]; Isotype: Mouse IgG2a, κ; Reactivity: Rat; Apps: FC; Size: 25 ug
Catalog No. 50-207-2525