Learn More
Custom Networks Inc APC Backplate Kit, 3x NEMA L5-20R Recepticals, for Smart-UPS Modular Ultra
SDP
Catalog No. NC3978261
Encompass_Preferred
The APC Backplate Kit with 3x NEMA L5-20R Receptacles is designed for use with Smart-UPS Modular Ultra models when the UPS installation is not hardwired. This kit provides ultra-high power density and on-line power protection capabilities for compatible UPS systems.
  • Backplate with 3 NEMA L5-20R outlets.
  • Provides 120V output.
  • Compatible with all Smart-UPS Modular Ultra models.
  • Designed for use when the UPS installation is not hardwired.
Catalog No. NC3978261 Supplier Custom Networks Inc Supplier No. SRYLPD1
The APC Backplate Kit with 3x NEMA L5-20R Receptacles is designed for use with Smart-UPS Modular Ultra models when the UPS installation is not hardwired. This kit provides ultra-high power density and on-line power protection capabilities for compatible UPS systems.
  • Backplate with 3 NEMA L5-20R outlets.
  • Provides 120V output.
  • Compatible with all Smart-UPS Modular Ultra models.
  • Designed for use when the UPS installation is not hardwired.