Learn More
Enterprise Technology Solutions APC Power Saving Back-UPS Pro 1500
SDP
Catalog No. 502097210
Encompass_Preferred
The Power-Saving Back-UPS Pro 1500 (230V) from APC provides power to your important computer components in the event of an electrical outage. The Uninterruptible Power Supply (UPS) contains a battery which continues to power computers, monitors and other components if the electricity is cut from them. An LCD status display provides information including remaining battery life, load capacity, and event notices. The battery is hot-swappable and easy to replace. An audible alarm alerts you of events like power outage and low battery.
Catalog No. 50-209-7210 Supplier Enterprise Technology Solutions Supplier No. BR1500G
The Power-Saving Back-UPS Pro 1500 (230V) from APC provides power to your important computer components in the event of an electrical outage. The Uninterruptible Power Supply (UPS) contains a battery which continues to power computers, monitors and other components if the electricity is cut from them. An LCD status display provides information including remaining battery life, load capacity, and event notices. The battery is hot-swappable and easy to replace. An audible alarm alerts you of events like power outage and low battery.