Learn More
Enterprise Technology Solutions APC Battery Back-UPS Pro BX1500M)â€
SDP
Catalog No. 502097240
Encompass_Preferred
Designed for electronics in your home or office, such as computers and related equipment, network devices (modem/router), POS devices, CCTV circuits, televisions, video game consoles, and more, the Battery Back-UPS Pro BX1500M from APC helps protect against surges, spikes, lightning, and outages. In the case of a power outage, it can provide up to 2.8 minutes of runtime at max load and 10.9 minutes at half load. It also provides an output capacity of 1500 VA / 900W, ten total NEMA 5-15R outlets (five battery backup & surge and five surge-only), an input voltage of 120 VAC at 60 Hz ± 3 Hz using its 6' cord with a NEMA 5-15P plug, a surge energy rating of 789 joules, and 1Gb of dataline protection. Its sealed, lead-acid battery is maintenance-free and can be replaced using an APCRBC124 replacement battery.
Catalog No. 50-209-7240 Supplier Enterprise Technology Solutions Supplier No. BX1500M
Designed for electronics in your home or office, such as computers and related equipment, network devices (modem/router), POS devices, CCTV circuits, televisions, video game consoles, and more, the Battery Back-UPS Pro BX1500M from APC helps protect against surges, spikes, lightning, and outages. In the case of a power outage, it can provide up to 2.8 minutes of runtime at max load and 10.9 minutes at half load. It also provides an output capacity of 1500 VA / 900W, ten total NEMA 5-15R outlets (five battery backup & surge and five surge-only), an input voltage of 120 VAC at 60 Hz ± 3 Hz using its 6' cord with a NEMA 5-15P plug, a surge energy rating of 789 joules, and 1Gb of dataline protection. Its sealed, lead-acid battery is maintenance-free and can be replaced using an APCRBC124 replacement battery.