Learn More
Abcam APC Conjugation Kit - Lightning-Link, 3 x 100UG
Catalog No. 502589117
Encompass_Preferred
APC conjugation / labeling in < 4 hrs with 30 secs hands-on time using APC Conjugation Kit ab201807. 100% antibody recovery. - Cited in over 50 publications - APC labeled antibody is ready for use in applications like WB, ELISA, and IHC, with no need for further purification - Compatible with most standard antibody formulations - Scalable;use the same conjugation method from 10µg-100mg of antibody - Use Lightning-Link conjugation with over 45 labels, including Alexa Fluor, DyLight, tandem dyes, enzymes, oligonucleotides, and gold nanoparticles
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-9117 Supplier Abcam Supplier No. AB2018073X100UG
APC conjugation / labeling in < 4 hrs with 30 secs hands-on time using APC Conjugation Kit ab201807. 100% antibody recovery. - Cited in over 50 publications - APC labeled antibody is ready for use in applications like WB, ELISA, and IHC, with no need for further purification - Compatible with most standard antibody formulations - Scalable;use the same conjugation method from 10µg-100mg of antibody - Use Lightning-Link conjugation with over 45 labels, including Alexa Fluor, DyLight, tandem dyes, enzymes, oligonucleotides, and gold nanoparticles
The product is subject to the following: Abcam Restricted Use Statement