Learn More
Biolegend APC/Cyanine7 anti-human IFN-γ
Supplier:  Biolegend 506524
Encompass_Preferred
APC/Cyanine7 anti-human IFN-γ [B27]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, African Green Monkey, Pigtailed Macaque, Sooty Mangabey; Apps: ICFC; Size: 100 tests
Catalog No. NC1899166