Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc APC-Cy7 aMu CD69 Anti Primary

Catalog No. 5000087290
Encompass_Preferred

Product Name: APC-Cyanine7 Anti-Mouse CD69 (H1.2F3)
Product Class: Antibody
Type: Primary
Product Name: APC-Cyanine7 Anti-Mouse CD69 (H1.2F3)
Cytek Catalog Number: 25-0691-U100
Marker: CD69
Target Species: Mouse
Clonality: Monoclonal
Clone: H1.2F3
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: APC-Cyanine7
Size: 100 μg

Catalog No. 50-000-87290 Supplier Cytek Biosciences Inc Supplier No. 250691U100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: APC-Cyanine7 Anti-Mouse CD69 (H1.2F3)
Product Class: Antibody
Type: Primary
Product Name: APC-Cyanine7 Anti-Mouse CD69 (H1.2F3)
Cytek Catalog Number: 25-0691-U100
Marker: CD69
Target Species: Mouse
Clonality: Monoclonal
Clone: H1.2F3
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: APC-Cyanine7
Size: 100 μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.