Learn More
Biolegend APC/Cyanine7 anti-human CD10
Supplier:  Biolegend 312212
Encompass_Preferred
APC/Cyanine7 anti-human CD10 [HI10a]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin monkey, Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-680