Learn More
Biolegend APC/Cyanine7 anti-human CD13
Supplier:  Biolegend 301710
Encompass_Preferred
APC/Cyanine7 anti-human CD13 [WM15]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cotton-topped Tamarin; Apps: FC; Size: 100 tests
Catalog No. 50-165-742