Learn More
Biolegend APC/Cyanine7 anti-human CD36
Supplier:  Biolegend 336213
Encompass_Preferred
APC/Cyanine7 anti-human CD36 [5-271]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-850