Learn More
Biolegend APC/Cyanine7 anti-human CD4
Supplier:  Biolegend 300517
Encompass_Preferred
APC/Cyanine7 anti-human CD4 [RPA-T4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. 50-165-591