Learn More
Biolegend APC/Cyanine7 anti-human IgM
Supplier:  Biolegend 314519
Encompass_Preferred
APC/Cyanine7 anti-human IgM [MHM-88]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-166-817