Learn More
Biolegend APC/Cyanine7 anti-human CD24
Supplier:  Biolegend 311131
Encompass_Preferred
APC/Cyanine7 anti-human CD24 [ML5]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee ; Apps: FC; Size: 25 tests
Catalog No. NC1070575