Learn More
Biolegend APC/Cyanine7 anti-human CD90 (Thy1)
Supplier:  Biolegend 328132
Encompass_Preferred
APC/Cyanine7 anti-human CD90 (Thy1) [5E10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Pigtailed Macaque, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 tests
Catalog No. NC1284366