Learn More
Biolegend APC/Cyanine7 anti-mouse CD103
Supplier:  Biolegend 121432
Encompass_Preferred
APC/Cyanine7 anti-mouse CD103 [2E7]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1115205