Learn More
Biolegend APC/Cyanine7 anti-mouse CD182 (CXCR2)
Catalog No. NC1739690
Encompass_Preferred
APC/Cyanine7 anti-mouse CD182 (CXCR2) [SA044G4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1739690 Supplier Biolegend Supplier No. 149313
APC/Cyanine7 anti-mouse CD182 (CXCR2) [SA044G4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug