Learn More
Biolegend APC/Cyanine7 anti-mouse CD16/32
Supplier:  Biolegend 101327
Encompass_Preferred
APC/Cyanine7 anti-mouse CD16/32 [93]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-555