Learn More
Biolegend APC/Cyanine7 anti-mouse CD25
Supplier:  Biolegend 102025
Encompass_Preferred
APC/Cyanine7 anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-631