Learn More
Biolegend APC/Cyanine7 anti-mouse CD49b (pan-NK cells)
Supplier:  Biolegend 108920
Encompass_Preferred
APC/Cyanine7 anti-mouse CD49b (pan-NK cells) [DX5]; Isotype: Rat IgM, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-402-983