Learn More
Biolegend APC/Cyanine7 anti-mouse CD86
Supplier:  Biolegend 105029
Encompass_Preferred
APC/Cyanine7 anti-mouse CD86 [GL-1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-163-016