Learn More
Biolegend APC/Cyanine7 anti-mouse TCR Vα2
Supplier:  Biolegend 127818
Encompass_Preferred
APC/Cyanine7 anti-mouse TCR Vα2 [B20.1]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-421