Learn More
Cytek Biosciences Inc APC-CY7 HU CD45 (HI30) 100T
Supplier:  Cytek Biosciences Inc 250459T100
Encompass_Preferred
APC-Cyanine7 Anti-Human CD45 (HI30) Isotype: Mouse IgG1, kappa Apps: FC Size: 100 tests
Catalog No. 50-105-4274