Learn More
Biolegend APC/Cyanine7 anti-human CD326 (Ep-CAM)
Supplier:  Biolegend 324245
Encompass_Preferred
APC/Cyanine7 anti-human CD326 (Ep-CAM) [9C4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-5653